Assay Details
Target Gene Details
Entrez Gene ID: | 104662 |
Gene Name: | TSR1 20S rRNA accumulation |
Gene Aliases: |
AU040765, AW550801, mKIAA1401 |
Location: |
Chr.11:74898080-74909340 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsr1 | NM_177325.3 | 6 | 1143 | NP_796299.2 |
AK030545.1 | 5 | 1049 | BAE20453.1 | |
AK129350.1 | 6 | 1097 | BAC98160.1 | |
AK134365.1 | 6 | 1127 | BAE22117.1 | |
AK154063.1 | 6 | 1130 | BAE32349.1 |
