Assay Details
Target Gene Details
Entrez Gene ID: | 104662 |
Gene Name: | TSR1 20S rRNA accumulation |
Gene Aliases: |
AU040765, AW550801, mKIAA1401 |
Location: |
Chr.11:74898080-74909340 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsr1 | NM_177325.3 | NP_796299.2 | ||
| AK030545.1 | BAE20453.1 | |||
| AK129350.1 | BAC98160.1 | |||
| AK154063.1 | BAE32349.1 | |||
| AK162686.1 | BAE37023.1 |

Genomic Map