Assay Details
Target Gene Details
Entrez Gene ID: | 108903 |
Gene Name: | tubulin-specific chaperone d |
Gene Aliases: |
2310057L06Rik, A030005L14Rik, mKIAA0988 |
Location: |
Chr.11:121450584-121617170 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbcd | NM_029878.3 | 13 | 1370 | NP_084154.1 |
| AK041464.1 | 13 | 1318 | BAC30951.1 | |
| AK050199.1 | 13 | 1373 | ||
| AK054343.1 | 13 | 1329 | ||
| AK185941.1 | 3 | 140 | ||
| BC059843.1 | 13 | 1290 | AAH59843.1 |
More Information
Set Membership: |
|

Genomic Map