Assay Details
Target Gene Details
Entrez Gene ID: | 72795 |
Gene Name: | tetratricopeptide repeat domain 19 |
Gene Aliases: |
2010204O13Rik, 2810460C24Rik, AI505442 |
Location: |
Chr.11:62281427-62316441 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc19 | NM_028360.2 | 9 | 878 | NP_082636.3 |
NM_029704.2 | 8 | 839 | NP_083980.2 | |
AK013368.1 | 9 | 831 | BAB28813.2 | |
AK034075.1 | 9 | 868 | BAC28572.1 | |
AK177481.1 | ||||
AK185142.1 | ||||
BC026866.1 | 7 | 522 | AAH26866.1 | |
BC037111.1 | 8 | 602 | AAH37111.1 |
