Assay Details
Target Gene Details
Entrez Gene ID: | 73936 |
Gene Name: | coiled-coil domain containing 175 |
Gene Aliases: |
4930403N07Rik |
Location: |
Chr.12:72100973-72185029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc175 | NM_028687.1 | NP_082963.1 |

Genomic Map