Assay Details
Target Gene Details
Entrez Gene ID: | 66601 |
Gene Name: | transmembrane and immunoglobulin domain containing 1 |
Gene Aliases: |
2010002A20Rik, Tmigd |
Location: |
Chr.11:76891246-76916587 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmigd1 | NM_025655.2 | 4 | 708 | NP_079931.1 |
AK008028.1 | 5 | 769 | BAB25419.1 | |
AK009118.1 | 4 | 708 | BAB26083.1 | |
BC022683.1 | 4 | 675 | AAH22683.1 | |
BC061082.1 | 4 | 698 | AAH61082.1 |
