Assay Details
Target Gene Details
Entrez Gene ID: | 66601 |
Gene Name: | transmembrane and immunoglobulin domain containing 1 |
Gene Aliases: |
2010002A20Rik, Tmigd |
Location: |
Chr.11:76891246-76916587 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmigd1 | NM_025655.2 | NP_079931.1 | ||
AK008028.1 | BAB25419.1 | |||
AK009118.1 | BAB26083.1 | |||
BC022683.1 | AAH22683.1 | |||
BC061082.1 | AAH61082.1 |
