Assay Details
Target Gene Details
Entrez Gene ID: | 217012 |
Gene Name: | unc-45 homolog B (C. elegans) |
Gene Aliases: |
AA445617, Cmya4, D230041A13Rik, Unc45 |
Location: |
Chr.11:82911253-82943406 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc45b | NM_178680.4 | NP_848795.3 | ||
| AF539793.1 | AAO13383.1 | |||
| AK052294.1 | BAC34921.1 | |||
| AK081821.1 | BAC38342.1 | |||
| AK084418.1 | BAC39178.1 | |||
| AK168968.1 | ||||
| BC084585.1 | AAH84585.1 |
More Information
Set Membership: |
|

Genomic Map