Assay Details
Target Gene Details
Entrez Gene ID: | 11491 |
Gene Name: | a disintegrin and metallopeptidase domain 17 |
Gene Aliases: |
CD156b, Tace |
Location: |
Chr.12:21323509-21373632 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam17 | NM_001277266.1 | NP_001264195.1 | ||
| NM_001291871.1 | NP_001278800.1 | |||
| NM_009615.6 | NP_033745.4 | |||
| NR_102380.1 | ||||
| NR_120376.1 | ||||
| AB021709.1 | BAA78578.1 | |||
| AJ007365.1 | CAA07480.1 | |||
| AK137693.1 | BAE23464.1 | |||
| AK139471.1 | BAE24023.1 | |||
| AK142441.1 | ||||
| AK144161.1 | BAE25740.1 | |||
| AK149288.1 | BAE28792.1 | |||
| AK149614.1 | BAE28991.1 | |||
| AK165739.1 | BAE38362.1 | |||
| BC094655.1 | AAH94655.1 | |||
| BC114999.1 | AAI15000.1 | |||
| BC138416.1 | ||||
| BC138420.1 | ||||
| BC145270.1 | ||||
| CA325381.1 | ||||
| U69613.1 | AAD09627.1 | |||
| U69614.1 | AAD09628.1 |
More Information
Set Membership: |
|

Genomic Map