Assay Details
Target Gene Details
Entrez Gene ID: | 21975 |
Gene Name: | topoisomerase (DNA) III alpha |
Gene Aliases: |
- |
Location: |
Chr.11:60739473-60777365 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top3a | NM_009410.2 | NP_033436.1 | ||
| AB006074.1 | BAA25662.1 | |||
| AK029987.1 | BAC26718.1 | |||
| AK037640.1 | BAE20515.1 | |||
| AK164443.1 | BAE37792.1 | |||
| AK170291.1 | BAE41691.1 | |||
| AK171286.1 | BAE42369.1 | |||
| BC099689.1 | AAH99689.1 |
More Information
Set Membership: |
|

Genomic Map