Assay Details
Target Gene Details
Entrez Gene ID: | 70834 |
Gene Name: | sperm associated antigen 9 |
Gene Aliases: |
3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1 |
Location: |
Chr.11:93996058-94126087 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag9 | NM_001199203.1 | NP_001186132.1 | ||
| NM_001199204.1 | NP_001186133.1 | |||
| NM_027569.2 | NP_081845.2 | |||
| AB047782.1 | BAD93176.1 | |||
| AF327451.1 | AAN61564.1 | |||
| AK030165.1 | ||||
| AK161623.1 | ||||
| AY823270.1 | AAX19462.1 | |||
| BB618351.1 | ||||
| FJ042496.1 |
More Information
Set Membership: |
|

Genomic Map