Assay Details
Target Gene Details
Entrez Gene ID: | 56427 |
Gene Name: | tubulin, delta 1 |
Gene Aliases: |
4930550G19Rik, Tubd |
Location: |
Chr.11:86544991-86567422 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubd1 | NM_001199045.1 | NP_001185974.1 | ||
| NM_001199046.1 | NP_001185975.1 | |||
| NM_001199047.1 | NP_001185976.1 | |||
| NM_019756.3 | NP_062730.1 | |||
| AB024329.1 | BAD16589.1 | |||
| AB031043.1 | BAD16591.1 | |||
| AF081568.1 | AAD52009.1 | |||
| AK016086.1 | ||||
| AK030214.1 | BAC26848.1 | |||
| AK169454.1 | BAE41182.1 | |||
| AK170304.1 | BAE41701.1 | |||
| AK171546.1 | BAE42519.1 | |||
| BC026465.1 | AAH26465.1 | |||
| BC132325.1 |
More Information
Set Membership: |
|

Genomic Map