Assay Details
Target Gene Details
Entrez Gene ID: | 56418 |
Gene Name: | YKT6 homolog (S. Cerevisiae) |
Gene Aliases: |
0610042I15Rik, 1810013M05Rik, AW105923 |
Location: |
Chr.11:5955758-5967780 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ykt6 | NM_019661.4 | NP_062635.2 | ||
AF076956.1 | AAC26834.1 | |||
AK002916.1 | BAB22455.1 | |||
AK007486.1 | BAB25062.1 | |||
AK147098.1 | BAE27674.1 | |||
AK150472.1 | BAE29589.1 | |||
AK159253.1 | BAE34934.1 | |||
AK211548.1 | ||||
BC006760.1 | AAH06760.1 |
