Assay Details
Target Gene Details
Entrez Gene ID: | 448850 |
Gene Name: | zinc finger, HIT type 3 |
Gene Aliases: |
Myohd1, Trip3 |
Location: |
Chr.11:84910950-84916356 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Znhit3 | NM_001005223.2 | NP_001005223.1 | ||
AK002888.1 | BAB22434.1 | |||
AK003721.1 | BAB22958.1 | |||
AK004259.1 | BAB23239.1 | |||
AK014350.1 | BAB29291.1 | |||
BC089554.1 | AAH89554.1 |
