Assay Details
Target Gene Details
Entrez Gene ID: | 217069 |
Gene Name: | tripartite motif-containing 25 |
Gene Aliases: |
AA960166, AL022677, EFP, Zfp147 |
Location: |
Chr.11:88999403-89020293 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim25 | NM_009546.2 | NP_033572.2 | ||
AK154533.1 | BAE32658.1 | |||
AK160898.1 | BAE36077.1 | |||
AK169562.1 | BAE41230.1 | |||
AK171744.1 | BAE42646.1 | |||
BC006908.1 | AAH06908.1 | |||
BC034276.1 | AAH34276.1 | |||
D63902.1 | BAA09941.1 |
