Assay Details
Target Gene Details
Entrez Gene ID: | 22187 |
Gene Name: | ubiquitin B |
Gene Aliases: |
AL033289, Rps27a, Uba52, Ubb2, Ubc |
Location: |
Chr.11:62551504-62553213 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubb | NM_011664.3 | NP_035794.1 | ||
AK003190.1 | BAB22630.1 | |||
AK007273.1 | BAB24930.1 | |||
AK010413.1 | BAB26919.1 | |||
AK010625.1 | BAB27071.1 | |||
AK012443.1 | BAB28242.1 | |||
AK013027.1 | BAB28606.1 | |||
AK013873.1 | BAB29028.1 | |||
AK132747.1 | BAE21330.1 | |||
AK160683.1 | ||||
AK165313.1 | ||||
BC066197.1 | AAH66197.1 | |||
X51703.1 | CAA35999.1 |
