Assay Details
Target Gene Details
Entrez Gene ID: | 66787 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
4933433P14Rik |
Location: |
Chr.12:105685352-105703058 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gskip | NM_178613.3 | NP_848728.2 | ||
AK020785.1 | ||||
AK042664.1 | BAC31325.1 | |||
AK079775.1 | BAC37748.1 | |||
AK167871.1 | ||||
AK168962.1 | BAE40767.1 | |||
AK171097.1 | BAE42246.1 | |||
BC031494.1 | AAH31494.1 |
