Assay Details
Target Gene Details
Entrez Gene ID: | 71275 |
Gene Name: | NADP+ dependent oxidoreductase domain containing 1 |
Gene Aliases: |
4933437F05Rik |
Location: |
Chr.12:87221123-87238774 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Noxred1 | NM_027744.1 | 6 | 797 | NP_082020.1 |
| AK017096.1 | 6 | 928 | BAB30594.1 | |
| BC125331.1 | 6 | 797 | ||
| BC125333.1 | 6 | 797 |
More Information
Set Membership: |
|

Genomic Map