Assay Details
Target Gene Details
Entrez Gene ID: | 69747 |
Gene Name: | zinc finger SWIM-type containing 7 |
Gene Aliases: |
2410012H22Rik |
Location: |
Chr.11:62267224-62281395 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zswim7 | NM_027198.1 | NP_081474.1 | ||
AK010472.1 | BAB26965.1 | |||
BC109343.2 | AAI09344.1 | |||
BQ554850.1 |
