Assay Details
Target Gene Details
Entrez Gene ID: | 217342 |
Gene Name: | ubiquitin-conjugating enzyme E2O |
Gene Aliases: |
9630022H21, B230113M03Rik, E2-230K, mKIAA1734 |
Location: |
Chr.11:116537740-116581476 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2o | NM_173755.3 | 8 | 1097 | NP_776116.2 |
AK129431.1 | 8 | 1097 | BAC98241.1 | |
BC059193.1 | 8 | 722 | AAH59193.1 | |
U20780.1 | 8 | 759 | AAA69916.1 |
