Assay Details
Target Gene Details
Entrez Gene ID: | 238055 |
Gene Name: | apolipoprotein B |
Gene Aliases: |
AI315052, Apo B-100, apob-100, apob-48 |
Location: |
Chr.12:7977633-8016839 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apob | NM_009693.2 | NP_033823.2 | ||
| BC038263.1 | AAH38263.1 | |||
| BC100607.1 | AAI00608.1 |
More Information
Set Membership: |
|

Genomic Map