Assay Details
Target Gene Details
Entrez Gene ID: | 79560 |
Gene Name: | ubiquitin-like domain containing CTD phosphatase 1 |
Gene Aliases: |
4930527B16Rik, 8430435I17Rik, BC002236 |
Location: |
Chr.11:44454571-44470548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ublcp1 | NM_024475.5 | NP_077795.2 | ||
AK043952.1 | BAC31711.1 | |||
AK050031.1 | BAC34039.1 | |||
AK147744.1 | BAE28110.1 | |||
AK153998.1 | ||||
BC002236.1 | AAH02236.1 | |||
BC056652.1 | AAH56652.1 | |||
BC083331.1 | AAH83331.1 | |||
BC085111.1 | AAH85111.1 | |||
BC106093.1 | AAI06094.1 |
