Assay Details
Target Gene Details
Entrez Gene ID: | 237898 |
Gene Name: | ubiquitin specific peptidase 32 |
Gene Aliases: |
2900074J03Rik, 6430526O11Rik, AW045245 |
Location: |
Chr.11:84984437-85140042 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp32 | NM_001029934.1 | 24 | 3039 | NP_001025105.1 |
| AK148221.1 | 24 | 3039 | ||
| BC051399.1 | 1 | 77 | AAH51399.1 | |
| BC058994.1 | 5 | 580 | AAH58994.1 |
More Information
Set Membership: |
|

Genomic Map