Assay Details
Target Gene Details
Entrez Gene ID: | 77706 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 5 |
Gene Aliases: |
9230106F14Rik |
Location: |
Chr.12:118867486-118966421 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb5 | NM_029961.2 | NP_084237.1 | ||
| AK139832.1 | BAE24153.1 | |||
| AY766239.1 | ||||
| JQ655148.1 |
More Information
Set Membership: |
|

Genomic Map