Assay Details
Target Gene Details
| Entrez Gene ID: | 21408 | 
| Gene Name: | zinc finger protein 354A | 
| Gene Aliases: | AW488485, Tcf17, Znf354a, kid1 | 
| Location: | Chr.11:51059244-51072799 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 1 - Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Zfp354a | NM_009329.3 | NP_033355.2 | ||
| AF184111.1 | AAF01033.1 | |||
| AK030469.1 | ||||
| BC050843.1 | AAH50843.1 | |||
| BC087540.1 | AAH87540.1 | |||
| L77247.1 | AAA96309.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic