Assay Details
Target Gene Details
Entrez Gene ID: | 278304 |
Gene Name: | zinc finger protein 385C |
Gene Aliases: |
A930006D11Rik, Znf385c |
Location: |
Chr.11:100626219-100692659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp385c | NM_177790.4 | NP_808458.2 | ||
| AK044299.1 | BAC31860.1 | |||
| BC116726.1 |

Genomic Map