Assay Details
Target Gene Details
Entrez Gene ID: | 67217 |
Gene Name: | L-3-hydroxyproline dehydratase (trans-) |
Gene Aliases: |
2810055F11Rik |
Location: |
Chr.12:72071660-72085433 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3hypdh | NM_026038.2 | NP_080314.1 | ||
| AK012954.1 | ||||
| AK018449.1 | BAB31217.1 | |||
| AK137154.1 | ||||
| BC004753.1 | AAH04753.1 |
More Information
Set Membership: |
|

Genomic Map