Assay Details
Target Gene Details
Entrez Gene ID: | 217578 |
Gene Name: | bromodomain adjacent to zinc finger domain 1A |
Gene Aliases: |
Acf1, B930060C03, BC065123, Gtl5, Wcrf180, cbp146 |
Location: |
Chr.12:54892600-55014371 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz1a | NM_013815.2 | NP_038843.2 | ||
AF033664.1 | AAC32043.1 | |||
AK135668.1 | BAE22607.1 | |||
AK166955.1 | BAE39138.1 |
