Assay Details
Target Gene Details
Entrez Gene ID: | 27386 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
4930423H22Rik, bHLHe12 |
Location: |
Chr.12:53248003-54072175 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npas3 | NM_013780.2 | NP_038808.2 | ||
| AF173871.1 | AAF14283.1 | |||
| BC120885.1 |
More Information
Set Membership: |
|

Genomic Map