Assay Details
Target Gene Details
Entrez Gene ID: | 217946 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
BC006933, JPO2 |
Location: |
Chr.12:117843475-117878712 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca7l | NM_146040.1 | 3 | 300 | NP_666152.1 |
AK149673.1 | 3 | 278 | BAE29017.1 | |
BC006933.1 | 3 | 300 | AAH06933.1 |
