Assay Details
Target Gene Details
Entrez Gene ID: | 70897 |
Gene Name: | family with sequence similarity 71, member D |
Gene Aliases: |
4921509E07Rik, 4930516C23Rik, GARI-L2 |
Location: |
Chr.12:78691488-78734519 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam71d | NM_027597.4 | NP_081873.2 | ||
NM_029069.1 | NP_083345.1 | |||
AK014851.1 | ||||
BC051149.1 | AAH51149.1 |
