Assay Details
Target Gene Details
Entrez Gene ID: | 67963 |
Gene Name: | Niemann-Pick type C2 |
Gene Aliases: |
2700012J19Rik, AA408070, AU045843, HE1 |
Location: |
Chr.12:84754559-84773112 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc2 | NM_023409.4 | NP_075898.1 | ||
AB021289.1 | BAA35183.1 | |||
AI391386.1 | ||||
AK008603.1 | BAB25771.1 | |||
AK009127.1 | BAB26090.1 | |||
AK012239.1 | ||||
AK149805.1 | ||||
AK151067.1 | BAE30083.1 | |||
AK151133.1 | BAE30141.1 | |||
AK152236.1 | BAE31061.1 | |||
AK153020.1 | BAE31654.1 | |||
AK153203.1 | BAE31803.1 | |||
AK154838.1 | BAE32866.1 | |||
AK158624.1 | BAE34585.1 | |||
AK159154.1 | BAE34861.1 | |||
AK165263.1 | BAE38111.1 | |||
AK167869.1 | BAE39885.1 | |||
AK170327.1 | BAE41721.1 | |||
BC003471.1 | AAH03471.1 | |||
BC007190.1 | AAH07190.1 |
