Assay Details
Target Gene Details
Entrez Gene ID: | 67963 |
Gene Name: | Niemann-Pick type C2 |
Gene Aliases: |
2700012J19Rik, AA408070, AU045843, HE1 |
Location: |
Chr.12:84754559-84773112 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc2 | NM_023409.4 | 3 | 524 | NP_075898.1 |
AB021289.1 | 3 | 279 | BAA35183.1 | |
AI391386.1 | ||||
AK008603.1 | 3 | 311 | BAB25771.1 | |
AK009127.1 | 3 | 341 | BAB26090.1 | |
AK012239.1 | 3 | 371 | ||
AK054441.1 | 1 | 1319 | ||
AK149805.1 | 3 | 313 | ||
AK151067.1 | 3 | 311 | BAE30083.1 | |
AK151080.1 | 1 | 1737 | ||
AK151133.1 | 3 | 312 | BAE30141.1 | |
AK152236.1 | 3 | 311 | BAE31061.1 | |
AK153020.1 | 3 | 311 | BAE31654.1 | |
AK153203.1 | 3 | 311 | BAE31803.1 | |
AK154838.1 | 3 | 311 | BAE32866.1 | |
AK158624.1 | 3 | 492 | BAE34585.1 | |
AK159154.1 | 3 | 358 | BAE34861.1 | |
AK165263.1 | 3 | 337 | BAE38111.1 | |
AK167869.1 | 3 | 312 | BAE39885.1 | |
AK170327.1 | 3 | 312 | BAE41721.1 | |
BC003471.1 | 3 | 326 | AAH03471.1 | |
BC007190.1 | 3 | 358 | AAH07190.1 |
