Assay Details
Target Gene Details
Entrez Gene ID: | 75302 |
Gene Name: | additional sex combs like 2 (Drosophila) |
Gene Aliases: |
4930556B16Rik, mKIAA1685 |
Location: |
Chr.12:3426857-3506852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asxl2 | NM_001270988.1 | NP_001257917.1 | ||
| NM_172421.5 | NP_766009.2 | |||
| AK016141.1 | ||||
| AK036839.1 | BAC29602.1 | |||
| AK080679.1 | BAC37978.2 | |||
| AK142135.1 | BAE24942.1 | |||
| AK148015.1 | ||||
| BC141140.1 |

Genomic Map