Assay Details
Target Gene Details
Entrez Gene ID: | 217946 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
BC006933, JPO2 |
Location: |
Chr.12:117843475-117878712 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca7l | NM_146040.1 | NP_666152.1 | ||
AK149673.1 | BAE29017.1 | |||
BC006933.1 | AAH06933.1 |
