Assay Details
Target Gene Details
Entrez Gene ID: | 52504 |
Gene Name: | centromere protein O |
Gene Aliases: |
2810429O05Rik, 8430427C03Rik, D12Ertd482e |
Location: |
Chr.12:4197058-4234294 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpo | NM_134046.5 | 6 | 1303 | NP_598807.2 |
AK013198.1 | BAB28707.1 | |||
AK029226.1 | 6 | 1304 | BAC26354.1 | |
AK032545.1 | 1 | 1167 | ||
AK036089.1 | 5 | 808 | BAC29301.1 | |
AK134266.1 | 6 | 975 | BAE22073.1 | |
BC023148.1 | 6 | 946 | AAH23148.1 | |
BC034399.1 | 6 | 962 | AAH34399.1 |
