Assay Details
Target Gene Details
Entrez Gene ID: | 14226 |
Gene Name: | FK506 binding protein 1b |
Gene Aliases: |
12.6kDa, AW494148, FKBP12.6, calstabin2 |
Location: |
Chr.12:4833174-4842463 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp1b | NM_016863.3 | 1 | 63 | NP_058559.3 |
| AB285017.1 | ||||
| AF060872.1 | AAC64923.1 | |||
| AK005201.1 | 1 | 64 | BAB23879.1 | |
| BC049596.1 | AAH49596.2 | |||
| BC061121.1 | AAH61121.1 | |||
| BE861051.1 | 1 | 78 |
More Information
Set Membership: |
|

Genomic Map