Assay Details
Target Gene Details
Entrez Gene ID: | 209334 |
Gene Name: | Gen homolog 1, endonuclease (Drosophila) |
Gene Aliases: |
5830483C08Rik |
Location: |
Chr.12:11239718-11266029 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gen1 | NM_177331.4 | 5 | 816 | NP_796305.3 |
| AK030995.1 | 5 | 826 | BAC27207.1 | |
| AK031077.1 | 5 | 795 | BAC27242.1 | |
| AK158255.1 | 4 | 639 | BAE34428.1 | |
| BC090653.1 | 5 | 816 | AAH90653.1 | |
| BC138157.1 | 5 | 703 | ||
| BC138158.1 | 5 | 703 |
More Information
Set Membership: |
|

Genomic Map