Assay Details
Target Gene Details
Entrez Gene ID: | 72258 |
Gene Name: | potassium channel, subfamily K, member 10 |
Gene Aliases: |
1700024D23Rik, 3010005K24Rik, Trek2 |
Location: |
Chr.12:98429432-98577940 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk10 | NM_029911.4 | NP_084187.2 | ||
| AK031904.1 | ||||
| AK036066.1 | BAC29295.1 | |||
| AK082153.1 | BAC38424.1 | |||
| BC132487.1 | ||||
| BC137869.1 | ||||
| DQ185134.1 | ABA28315.1 |
More Information
Set Membership: |
|

Genomic Map