Assay Details
Target Gene Details
Entrez Gene ID: | 15251 |
Gene Name: | hypoxia inducible factor 1, alpha subunit |
Gene Aliases: |
AA959795, HIF1alpha, MOP1, bHLHe78 |
Location: |
Chr.12:73901329-73947530 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hif1a | NM_010431.2 | 11 | 2029 | NP_034561.2 |
| AF003695.1 | 11 | 1863 | AAC53455.1 | |
| AK034087.1 | 11 | 1995 | BAC28578.1 | |
| AK076395.1 | 9 | 1317 | BAC36320.1 | |
| AK150367.1 | 11 | 1896 | BAE29500.1 | |
| BC026139.1 | 11 | 1881 | AAH26139.1 | |
| X95580.1 | CAA64833.1 |
More Information
Set Membership: |
|

Genomic Map