Assay Details
Target Gene Details
Entrez Gene ID: | 211914 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
6530401G17Rik, Ddef2, Gm1523, Gm592, PAG3, PAP |
Location: |
Chr.12:21111713-21270171 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asap2 | NM_001004364.2 | 16 | 1795 | NP_001004364.2 |
| NM_001098168.1 | 17 | 1804 | NP_001091637.1 | |
| NM_001135192.1 | 17 | 1795 | NP_001128664.1 | |
| AK147622.1 | 17 | 1803 | BAE28030.1 | |
| AK147656.1 | 17 | 1750 | BAE28052.1 | |
| AK172944.1 | 17 | 1616 | BAD32222.1 | |
| BC096022.1 | 17 | 1572 | AAH96022.1 |

Genomic Map