Assay Details
Target Gene Details
Entrez Gene ID: | 268566 |
Gene Name: | gephyrin |
Gene Aliases: |
5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph |
Location: |
Chr.12:78224153-78684772 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gphn | NM_145965.2 | NP_666077.2 | ||
| NM_172952.3 | NP_766540.2 | |||
| AK082353.1 | BAC38476.1 | |||
| AK144569.1 | ||||
| AK183551.1 | ||||
| BC127058.1 |
More Information
Set Membership: |
|

Genomic Map