Assay Details
Target Gene Details
Entrez Gene ID: | 338320 |
Gene Name: | melanoma inhibitory activity 2 |
Gene Aliases: |
- |
Location: |
Chr.12:59095800-59109130 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mia2 | NM_177321.2 | NP_796295.1 | ||
| AF390177.2 | AAL26992.2 |
More Information
Set Membership: |
|

Genomic Map