Assay Details
Target Gene Details
Entrez Gene ID: | 104721 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 1 |
Gene Aliases: |
AA409185, DBP-RB |
Location: |
Chr.12:13219307-13249174 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx1 | NM_134040.1 | 8 | 492 | NP_598801.1 |
AK028341.1 | 8 | 489 | BAC25893.1 | |
AK132157.1 | 8 | 532 | BAE21002.1 | |
AK153335.1 | 8 | 516 | BAE31914.1 | |
AK160982.1 | 8 | 493 | BAE36130.1 | |
AK166146.1 | 8 | 472 | ||
AK217868.1 | 2 | 108 | ||
BC008570.1 | 8 | 470 | AAH08570.1 | |
BC010624.1 | 8 | 492 | AAH10624.1 |
