Assay Details
Target Gene Details
Entrez Gene ID: | 14026 |
Gene Name: | Ena-vasodilator stimulated phosphoprotein |
Gene Aliases: |
AI528774 |
Location: |
Chr.12:108554720-108688516 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Evl | NM_001163394.1 | NP_001156866.1 | ||
| NM_001163395.1 | NP_001156867.1 | |||
| NM_001163396.2 | NP_001156868.1 | |||
| NM_007965.3 | NP_031991.3 | |||
| AF279662.1 | AAG23653.1 | |||
| AK134563.1 | ||||
| AK137823.1 | ||||
| AK151633.1 | ||||
| BC059810.1 | AAH59810.1 | |||
| U72519.1 | AAC52862.1 |
More Information
Set Membership: |
|

Genomic Map