Assay Details
Target Gene Details
Entrez Gene ID: | 16981 |
Gene Name: | leucine rich repeat protein 3, neuronal |
Gene Aliases: |
NLRR-3 |
Location: |
Chr.12:41451668-41486057 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrn3 | NM_001271708.1 | 1 | 397 | NP_001258637.1 |
| NM_001271709.1 | 1 | 397 | NP_001258638.1 | |
| NM_010733.3 | 1 | 397 | NP_034863.1 | |
| AK036316.1 | 1 | 294 | BAC29381.1 | |
| BC069041.1 | 1 | 91 | AAH69041.1 | |
| CJ058856.1 | 1 | 294 | ||
| CJ061095.1 | 1 | 294 | ||
| D49802.1 | 1 | 308 | BAA08622.1 |
More Information
Set Membership: |
|

Genomic Map