Assay Details
Target Gene Details
Entrez Gene ID: | 217893 |
Gene Name: | phosphofurin acidic cluster sorting protein 2 |
Gene Aliases: |
6720425G15Rik, AW208651, AW489976, E230011J22, Pacs1l, mKIAA0602 |
Location: |
Chr.12:113014507-113074401 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pacs2 | NM_001081170.1 | 23 | 2555 | NP_001074639.1 |
NM_001291444.1 | 23 | 2561 | NP_001278373.1 | |
NM_001291445.1 | 22 | 2465 | NP_001278374.1 | |
AK030003.1 | 18 | 1924 | BAE20449.1 | |
AK036581.1 | 22 | 2441 | BAE20504.1 | |
AK054000.1 | 23 | 2536 | ||
AK122326.1 | 21 | 2244 | BAC65608.1 | |
AK193976.1 | 2 | 282 | ||
AK200620.1 | ||||
BC043302.1 | 4 | 472 | AAH43302.1 | |
BC059076.1 | 8 | 1125 | AAH59076.1 |
