Assay Details
Target Gene Details
Entrez Gene ID: | 16413 |
Gene Name: | integrin beta 1 binding protein 1 |
Gene Aliases: |
AI449260, AU019480, Icap1 |
Location: |
Chr.12:21267246-21286279 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb1bp1 | NM_008403.4 | 6 | 649 | NP_032429.1 |
AJ001373.1 | 6 | 631 | CAA04706.1 | |
AK002786.1 | 6 | 606 | BAB22357.1 | |
AK004172.1 | 6 | 605 | BAB23206.1 | |
AK007635.1 | 6 | 607 | BAB25151.1 | |
AK075577.1 | 6 | 650 | BAC35832.1 | |
AK089937.1 | 6 | 763 | BAC41007.1 | |
AK147929.1 | 6 | 598 | BAE28236.1 | |
AK210343.1 | ||||
AK218590.1 | ||||
BC028772.1 | 5 | 485 | AAH28772.1 |
