Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | 19 | 4871 | NP_001074922.1 |
| NM_001286079.2 | 20 | 5053 | NP_001273008.1 | |
| NM_001286080.2 | 21 | 5100 | NP_001273009.1 | |
| NM_008697.4 | 21 | 5100 | NP_032723.2 | |
| NR_104397.2 | 21 | 5100 | ||
| NR_104398.2 | 19 | 2944 | ||
| AK129394.1 | 21 | 4990 | BAC98204.1 | |
| AK134860.1 | 19 | 2944 | ||
| AK147187.1 | 4 | 1856 | ||
| AK162150.1 | 20 | 2924 | ||
| AK167264.1 | 17 | 3966 | ||
| AY515727.1 | 19 | 4850 | AAS87211.1 | |
| BC137789.1 | 20 | 4938 | ||
| BC144913.1 | 21 | 4985 | ||
| U40342.1 | 21 | 5007 | AAA83234.1 |

Genomic Map