Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | 24 | 5570 | NP_001074922.1 |
| NM_001286079.2 | 25 | 5752 | NP_001273008.1 | |
| NM_001286080.2 | 26 | 5799 | NP_001273009.1 | |
| NM_008697.4 | 26 | 5799 | NP_032723.2 | |
| NR_104397.2 | 26 | 5799 | ||
| NR_104398.2 | 24 | 3643 | ||
| AK089140.1 | 1 | 762 | ||
| AK129394.1 | 26 | 5689 | BAC98204.1 | |
| AK134860.1 | 24 | 3643 | ||
| AK147187.1 | 9 | 2555 | ||
| AK167264.1 | 22 | 4665 | ||
| AY515727.1 | 24 | 5549 | AAS87211.1 | |
| BC137789.1 | 25 | 5637 | ||
| BC144913.1 | 26 | 5684 | ||
| U40342.1 | 26 | 5706 | AAA83234.1 |

Genomic Map